Assay Details
Target Gene Details
Entrez Gene ID: | 25901 |
Gene Name: | coiled-coil domain containing 28A |
Gene Aliases: |
C6orf80, CCRL1AP |
Location: |
Chr.6:138773517-138793319 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC28A | NM_015439.2 | NP_056254.1 | ||
| XM_011535728.2 | XP_011534030.1 | |||
| AJ420568.1 | ||||
| AK300672.1 | ||||
| AL050197.1 | CAB43315.1 | |||
| AY167571.1 | AAN87344.1 | |||
| BC004464.1 | AAH04464.1 | |||
| BC013019.2 | ||||
| BC080566.1 | AAH80566.1 | |||
| BC108717.1 | AAI08718.1 |
More Information
Set Membership: |
|

Genomic Map