Assay Details
Target Gene Details
Entrez Gene ID: | 5754 |
Gene Name: | protein tyrosine kinase 7 (inactive) |
Gene Aliases: |
CCK-4, CCK4 |
Location: |
Chr.6:43076268-43161720 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTK7 | NM_001270398.1 | NP_001257327.1 | ||
| NM_002821.4 | NP_002812.2 | |||
| NM_152880.3 | NP_690619.1 | |||
| NM_152881.3 | NP_690620.1 | |||
| NM_152882.3 | NP_690621.1 | |||
| NR_072997.1 | ||||
| NR_072998.1 | ||||
| XM_011514765.2 | XP_011513067.1 | |||
| XM_011514766.2 | XP_011513068.1 | |||
| AB209350.1 | BAD92587.1 | |||
| AF531868.1 | AAN04862.1 | |||
| AF531869.1 | AAN04863.1 | |||
| AF531870.1 | AAN04864.1 | |||
| AF531871.1 | AAN04865.1 | |||
| AF531872.1 | AAN04866.1 | |||
| AK291016.1 | ||||
| AK292589.1 | ||||
| AK296931.1 | ||||
| AK296953.1 | ||||
| BC046109.2 | AAH46109.1 | |||
| BC071557.1 | AAH71557.1 | |||
| CA389442.1 | ||||
| CN266652.1 | ||||
| U33635.1 | AAA87565.1 | |||
| U40271.2 | AAC50484.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950169 | Chr.6:43068163 - 43080562 on Build GRCh38 | Deletion |
|
| nsv602992 | Chr.6:42948182 - 43110718 on Build GRCh38 | Loss |
|
| nsv1017398 | Chr.6:42659705 - 43400998 on Build GRCh38 | Gain |
|
| nsv437509 | Chr.6:43070325 - 43116036 on Build GRCh38 | Loss |
|
| dgv10640n54 | Chr.6:43002769 - 43128461 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map