Assay Details
Target Gene Details
Entrez Gene ID: | 135138 |
Gene Name: | PARK2 coregulated |
Gene Aliases: |
GLUP, HAK005771, PARK2CRG |
Location: |
Chr.6:162726870-163315492 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Target Gene Details
Entrez Gene ID: | 101929239 |
Gene Name: | PACRG antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.6:163042980-163054161 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PACRG-AS2 | NR_110871.1 | |||
| AK058177.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv605159 | Chr.6:162379124 - 163058475 on Build GRCh38 | Loss |
|
| nsv1021433 | Chr.6:162954728 - 163088834 on Build GRCh38 | Loss |
|
| nsv605187 | Chr.6:163027718 - 163075257 on Build GRCh38 | Loss |
|
| nsv1026490 | Chr.6:162712401 - 163284676 on Build GRCh38 | Gain |
|
| nsv1030116 | Chr.6:162852675 - 163104646 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map