Assay Details
Target Gene Details
Entrez Gene ID: | 80380 |
Gene Name: | programmed cell death 1 ligand 2 |
Gene Aliases: |
B7DC, Btdc, CD273, PD-L2, PDCD1L2, PDL2, bA574F11.2 |
Location: |
Chr.9:5510438-5571282 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDCD1LG2 | NM_025239.3 | NP_079515.2 | ||
| XM_005251600.3 | XP_005251657.1 | |||
| AF329193.1 | AAK31105.1 | |||
| AF344424.1 | AAK15370.1 | |||
| AY254343.1 | AAP13471.1 | |||
| AY271901.1 | AAP49000.1 | |||
| AY271902.1 | AAP49001.1 | |||
| BC074766.2 | AAH74766.1 | |||
| BC113678.1 | ||||
| BC113680.1 | ||||
| DQ336699.1 | ABC59616.1 | |||
| EF444806.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509292 | Chr.9:5469291 - 5546768 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map