Assay Details
Target Gene Details
Entrez Gene ID: | 256691 |
Gene Name: | MAM domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.9:70043519-70226972 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAMDC2 | NM_153267.4 | NP_694999.3 | ||
| NR_125850.1 | ||||
| XM_011518471.1 | XP_011516773.1 | |||
| XM_011518475.2 | XP_011516777.1 | |||
| XM_017014587.1 | XP_016870076.1 | |||
| AK304040.1 | ||||
| AK304531.1 | ||||
| BC016383.1 | AAH16383.1 | |||
| BC063634.1 | AAH63634.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831613 | Chr.9:69971142 - 70125145 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs200860526] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map