Assay Details
Target Gene Details
Entrez Gene ID: | 158219 |
Gene Name: | tetratricopeptide repeat domain 39B |
Gene Aliases: |
C9orf52 |
Location: |
Chr.9:15167204-15307360 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC39B |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3619769 | Chr.9:15166239 - 15337990 on Build GRCh38 | Gain |
|
| nsv831519 | Chr.9:15018162 - 15209129 on Build GRCh38 | Loss |
|
| nsv524381 | Chr.9:15165034 - 15202419 on Build GRCh38 | Gain |
|
| esv3619766 | Chr.9:15163388 - 15206006 on Build GRCh38 | Gain |
|
| esv3619763 | Chr.9:15140390 - 15207671 on Build GRCh38 | Gain |
|

Genomic Map