Assay Details
Target Gene Details
Entrez Gene ID: | 340485 |
Gene Name: | alkaline ceramidase 2 |
Gene Aliases: |
ALKCDase2, ASAH3L |
Location: |
Chr.9:19408927-19455173 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACER2 | NM_001010887.2 | NP_001010887.2 | ||
| XM_005251448.3 | XP_005251505.1 | |||
| XM_011517858.1 | XP_011516160.1 | |||
| AF370405.1 | AAQ15241.1 | |||
| AK123581.1 | ||||
| AY312516.1 | AAQ85132.1 | |||
| BC092487.1 | AAH92487.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1025392 | Chr.9:19381646 - 19478974 on Build GRCh38 | Gain |
|
| nsv613731 | Chr.9:19400353 - 19448475 on Build GRCh38 | Loss |
|
| esv3619907 | Chr.9:19322693 - 19441414 on Build GRCh38 | Gain |
|
| nsv613730 | Chr.9:19281101 - 19516865 on Build GRCh38 | Gain |
|
| esv3619911 | Chr.9:19379247 - 19517379 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map