Assay Details
Target Gene Details
Entrez Gene ID: | 1759 |
Gene Name: | dynamin 1 |
Gene Aliases: |
DNM, EIEE31 |
Location: |
Chr.9:128203355-128255248 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Entrez Gene ID: | 1759 |
Gene Name: | dynamin 1 |
Gene Aliases: |
DNM, EIEE31 |
Location: |
Chr.9:128203355-128255248 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Set Membership: |
|