Assay Details
Target Gene Details
Entrez Gene ID: | 153090 |
Gene Name: | DAB2 interacting protein |
Gene Aliases: |
AF9Q34, AIP-1, AIP1, DIP1/2 |
Location: |
Chr.9:121566883-121785530 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DAB2IP | NM_032552.3 | NP_115941.2 | ||
| XM_005251721.1 | XP_005251778.1 | |||
| AK096391.1 | ||||
| BM545676.1 | ||||
| BM546615.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471468 | Chr.9:121564168 - 121598707 on Build GRCh38 | Loss |
|
| nsv520039 | Chr.9:121527783 - 121600223 on Build GRCh38 | Loss |
|
| nsv1037310 | Chr.9:121522791 - 121618643 on Build GRCh38 | Gain |
|
| nsv831712 | Chr.9:121503703 - 121659281 on Build GRCh38 | Loss |
|
| esv2663571 | Chr.9:121570764 - 121576559 on Build GRCh38 | Deletion |
|
| esv3621665 | Chr.9:121570762 - 121576516 on Build GRCh38 | Loss |
|
| esv29959 | Chr.9:121569278 - 121619614 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map