Assay Details
Target Gene Details
Entrez Gene ID: | 158135 |
Gene Name: | tubulin tyrosine ligase like 11 |
Gene Aliases: |
C9orf20, bA244O19.1 |
Location: |
Chr.9:121815674-122093606 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTLL11 | NM_001139442.1 | NP_001132914.1 | ||
| XM_005251728.2 | XP_005251785.1 | |||
| XM_005251729.4 | XP_005251786.1 | |||
| AK125178.1 | BAC86073.1 |
More Information
Set Membership: |
|

Genomic Map