Assay Details
Target Gene Details
Entrez Gene ID: | 254956 |
Gene Name: | MORN repeat containing 5 |
Gene Aliases: |
C9orf113, C9orf18 |
Location: |
Chr.9:122159661-122200088 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MORN5 | NM_001286828.1 | NP_001273757.1 | ||
| NM_198469.3 | NP_940871.2 | |||
| XM_005251877.4 | 4 | 1615 | XP_005251934.1 | |
| XM_005251878.3 | XP_005251935.1 | |||
| XM_006717034.3 | XP_006717097.1 | |||
| XM_011518457.2 | XP_011516759.1 | |||
| XM_011518458.1 | XP_011516760.1 | |||
| AI026684.1 | ||||
| AI560944.1 | ||||
| AK128877.1 | BAC87653.1 | |||
| AK302081.1 | ||||
| BC133690.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6702 | Chr.9:122151820 - 122184094 on Build GRCh38 | Insertion |
|
| nsv831715 | Chr.9:122066612 - 122226720 on Build GRCh38 | Gain |
|

Genomic Map