Assay Details
Target Gene Details
Entrez Gene ID: | 23732 |
Gene Name: | ferric chelate reductase 1 like |
Gene Aliases: |
C9orf4, CG-6, CG6, EIEE37 |
Location: |
Chr.9:109136188-109167291 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FRRS1L | NM_014334.2 | NP_055149.2 | ||
| XM_011518453.2 | XP_011516755.1 | |||
| XM_011518454.2 | XP_011516756.1 | |||
| AF155065.1 | AAF28159.1 | |||
| BF514667.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3621430 | Chr.9:109143603 - 109144777 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map