Assay Details
Target Gene Details
Entrez Gene ID: | 2934 |
Gene Name: | gelsolin |
Gene Aliases: |
ADF, AGEL |
Location: |
Chr.9:121201082-121332844 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GSN | XM_011518591.2 | XP_011516893.1 | ||
| XM_017014643.1 | XP_016870132.1 | |||
| XM_017014644.1 | XP_016870133.1 | |||
| XM_017014647.1 | XP_016870136.1 | |||
| AK125819.1 |
More Information
Set Membership: |
|

Genomic Map