Assay Details
Target Gene Details
Entrez Gene ID: | 2516 |
Gene Name: | nuclear receptor subfamily 5 group A member 1 |
Gene Aliases: |
AD4BP, ELP, FTZ1, FTZF1, POF7, SF-1, SF1, SPGF8, SRXY3, hSF-1 |
Location: |
Chr.9:124481234-124507420 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NR5A1 | NM_004959.4 | NP_004950.2 | ||
| XM_005251871.3 | XP_005251928.1 | |||
| XM_011518455.2 | XP_011516757.1 | |||
| AB307718.1 | ||||
| AK090858.1 | ||||
| BC032501.1 | AAH32501.1 | |||
| HQ692856.1 | ||||
| HQ709184.1 | ||||
| U76388.1 | AAB53105.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3621722 | Chr.9:124378965 - 124497390 on Build GRCh38 | Gain |
|
| nsv950942 | Chr.9:124466322 - 124513521 on Build GRCh38 | Deletion |
|
| nsv526209 | Chr.9:124470928 - 124531906 on Build GRCh38 | Gain |
|
| nsv517372 | Chr.9:124467839 - 124512572 on Build GRCh38 | Loss |
|
| nsv615291 | Chr.9:124464413 - 124557925 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76399357] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map