Assay Details
Target Gene Details
Entrez Gene ID: | 84628 |
Gene Name: | netrin G2 |
Gene Aliases: |
LHLL9381, Lmnt2, NTNG1, bA479K20.1 |
Location: |
Chr.9:132161234-132244534 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509328 | Chr.9:132084017 - 132270727 on Build GRCh38 | Insertion |
|
| nsv615572 | Chr.9:132183292 - 132251071 on Build GRCh38 | Gain |
|
| nsv1049356 | Chr.9:132170367 - 132210675 on Build GRCh38 | Gain |
|
| nsv1035219 | Chr.9:132147299 - 132243197 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map