Assay Details
Target Gene Details
Entrez Gene ID: | 256691 |
Gene Name: | MAM domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.9:70043519-70226972 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAMDC2 | NM_153267.4 | NP_694999.3 | ||
| NR_125850.1 | ||||
| XM_011518471.1 | XP_011516773.1 | |||
| XM_011518475.2 | XP_011516777.1 | |||
| XM_017014587.1 | XP_016870076.1 | |||
| AK304040.1 | ||||
| AK304531.1 | ||||
| BC015417.2 | AAH15417.1 | |||
| BC016383.1 | AAH16383.1 | |||
| BC063634.1 | AAH63634.1 |
Target Gene Details
Entrez Gene ID: | 100507244 |
Gene Name: | MAMDC2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.9:70153134-70175888 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAMDC2-AS1 | NR_040076.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047862 | Chr.9:70131959 - 70523371 on Build GRCh38 | Gain |
|

Genomic Map