Assay Details
Target Gene Details
Entrez Gene ID: | 158219 |
Gene Name: | tetratricopeptide repeat domain 39B |
Gene Aliases: |
C9orf52 |
Location: |
Chr.9:15167204-15307360 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC39B | NM_001168339.1 | 20 | 2983 | NP_001161811.1 |
| NM_001168340.1 | 19 | 2950 | NP_001161812.1 | |
| NM_001168341.1 | 18 | 2782 | NP_001161813.1 | |
| NM_001168342.1 | 18 | 2864 | NP_001161814.1 | |
| NM_152574.2 | 20 | 2989 | NP_689787.2 | |
| XM_011517732.2 | 19 | 2786 | XP_011516034.1 | |
| XM_017014310.1 | 22 | 3352 | XP_016869799.1 | |
| XM_017014311.1 | 21 | 3284 | XP_016869800.1 | |
| XM_017014312.1 | 19 | 2854 | XP_016869801.1 | |
| AI473911.1 | 1 | 187 | ||
| BC142985.1 | 20 | 2962 | ||
| BX105862.1 | 1 | 224 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3619769 | Chr.9:15166239 - 15337990 on Build GRCh38 | Gain |
|
| nsv831519 | Chr.9:15018162 - 15209129 on Build GRCh38 | Loss |
|
| nsv524381 | Chr.9:15165034 - 15202419 on Build GRCh38 | Gain |
|
| esv3619766 | Chr.9:15163388 - 15206006 on Build GRCh38 | Gain |
|
| esv3619763 | Chr.9:15140390 - 15207671 on Build GRCh38 | Gain |
|

Genomic Map