Assay Details
Target Gene Details
Entrez Gene ID: | 158135 |
Gene Name: | tubulin tyrosine ligase like 11 |
Gene Aliases: |
C9orf20, bA244O19.1 |
Location: |
Chr.9:121815674-122093606 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTLL11 | XM_005251728.2 | 8 | 6843 | XP_005251785.1 |
| XM_005251729.4 | 9 | 7920 | XP_005251786.1 | |
| AK123875.1 | 1 | 853 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525090 | Chr.9:121811226 - 121819482 on Build GRCh38 | Loss |
|
| nsv1053662 | Chr.9:121750763 - 121831165 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map