Assay Details
Target Gene Details
Entrez Gene ID: | 79048 |
Gene Name: | SECIS binding protein 2 |
Gene Aliases: |
SBP2 |
Location: |
Chr.9:89318473-89367117 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SECISBP2 | NM_001282688.1 | NP_001269617.1 | ||
| NM_001282689.1 | NP_001269618.1 | |||
| NM_001282690.1 | NP_001269619.1 | |||
| NM_024077.4 | NP_076982.3 | |||
| XM_005252196.2 | XP_005252253.1 | |||
| XM_006717282.2 | XP_006717345.1 | |||
| XM_011519000.2 | XP_011517302.1 | |||
| XM_011519001.1 | XP_011517303.1 | |||
| XM_011519002.1 | XP_011517304.1 | |||
| XM_011519003.1 | XP_011517305.1 | |||
| XM_017015122.1 | XP_016870611.1 | |||
| XM_017015123.1 | XP_016870612.1 | |||
| XM_017015124.1 | XP_016870613.1 | |||
| XM_017015125.1 | XP_016870614.1 | |||
| AB208940.1 | BAD92177.1 | |||
| AF380995.1 | AAK57518.1 | |||
| AK090608.1 | ||||
| AK290182.1 | ||||
| AK299161.1 | ||||
| AK302852.1 | ||||
| AL136881.1 | CAB66815.1 | |||
| BC001189.1 | AAH01189.1 | |||
| BC036109.1 | AAH36109.1 | |||
| BC099900.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3621014 | Chr.9:88877891 - 89641453 on Build GRCh38 | Gain |
|
| nsv1041327 | Chr.9:89298100 - 89438209 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map