Assay Details
Target Gene Details
Entrez Gene ID: | 79109 |
Gene Name: | mitogen-activated protein kinase associated protein 1 |
Gene Aliases: |
JC310, MIP1, SIN1, SIN1b, SIN1g |
Location: |
Chr.9:125437394-125707234 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAPKAP1 | NM_001006617.1 | NP_001006618.1 | ||
| NM_001006619.1 | NP_001006620.1 | |||
| NM_001006620.1 | NP_001006621.1 | |||
| NM_001006621.1 | NP_001006622.1 | |||
| NM_024117.3 | NP_077022.1 | |||
| XM_011519005.1 | XP_011517307.1 | |||
| XM_011519006.2 | XP_011517308.1 | |||
| XM_011519007.1 | XP_011517309.1 | |||
| XM_011519010.2 | XP_011517312.1 | |||
| XM_017015126.1 | XP_016870615.1 | |||
| XM_017015127.1 | XP_016870616.1 | |||
| AK290060.1 | ||||
| AK295364.1 | ||||
| AK298841.1 | ||||
| AK310361.1 | ||||
| AK316149.1 | ||||
| AL833042.1 | CAH56311.1 | |||
| AY524429.1 | AAS90839.1 | |||
| AY524431.1 | AAS90841.1 | |||
| AY524432.1 | AAS90842.1 | |||
| AY633624.1 | AAT46478.1 | |||
| AY633625.1 | AAT46479.1 | |||
| AY633626.1 | AAT46480.1 | |||
| BC002326.2 | AAH02326.1 | |||
| BC003044.1 | AAH03044.1 | |||
| M37191.1 | AAA36551.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831722 | Chr.9:125302099 - 125476079 on Build GRCh38 | Gain |
|
| nsv1046626 | Chr.9:125359313 - 125470470 on Build GRCh38 | Gain |
|
| nsv515979 | Chr.9:125388505 - 125465747 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114036737] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map