Assay Details
Target Gene Details
Entrez Gene ID: | 10044 |
Gene Name: | SH2 domain containing 3C |
Gene Aliases: |
CHAT, NSP3, PRO34088, SHEP1 |
Location: |
Chr.9:127738317-127778769 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SH2D3C | NM_001142531.1 | NP_001136003.1 | ||
| NM_001142532.1 | NP_001136004.1 | |||
| NM_001142533.1 | NP_001136005.1 | |||
| NM_001142534.1 | NP_001136006.1 | |||
| NM_001252334.1 | NP_001239263.1 | |||
| NM_005489.3 | NP_005480.2 | |||
| NM_170600.2 | NP_733745.1 | |||
| XM_005251639.1 | XP_005251696.1 | |||
| XM_011518114.1 | XP_011516416.1 | |||
| XM_011518115.1 | XP_011516417.1 | |||
| XM_011518117.2 | XP_011516419.1 | |||
| XM_017014174.1 | XP_016869663.1 | |||
| AF124251.1 | AAD28246.1 | |||
| AK056068.1 | ||||
| AK096983.1 | ||||
| AK128287.1 | ||||
| AK291393.1 | ||||
| AK292041.1 | ||||
| AK292718.1 | ||||
| AK294386.1 | ||||
| AY358089.1 | AAQ88456.1 | |||
| AY358440.1 | AAQ89948.1 | |||
| BC027962.1 | AAH27962.1 | |||
| BC032365.1 | AAH32365.1 | |||
| BX647905.1 | CAI46101.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv12879n54 | Chr.9:127731234 - 127824409 on Build GRCh38 | Loss |
|
| nsv825094 | Chr.9:127742027 - 127802620 on Build GRCh38 | Gain |
|
| nsv615369 | Chr.9:127707924 - 127774438 on Build GRCh38 | Loss |
|
| nsv471323 | Chr.9:127727464 - 127845239 on Build GRCh38 | Loss |
|
| nsv466573 | Chr.9:127735841 - 127787855 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map