Assay Details
Target Gene Details
Entrez Gene ID: | 883 |
Gene Name: | kynurenine aminotransferase 1 |
Gene Aliases: |
CCBL1, GTK, KAT1, KATI |
Location: |
Chr.9:128832942-128882551 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KYAT1 | NM_001122671.1 | NP_001116143.1 | ||
| NM_001122672.1 | NP_001116144.1 | |||
| NM_001287390.1 | 3 | 460 | NP_001274319.1 | |
| NM_004059.4 | NP_004050.3 | |||
| NR_109829.1 | ||||
| XM_006717311.3 | XP_006717374.1 | |||
| XM_011519165.2 | 3 | 431 | XP_011517467.1 | |
| XM_011519167.2 | XP_011517469.2 | |||
| XM_011519169.1 | XP_011517471.1 | |||
| XM_011519170.2 | XP_011517472.1 | |||
| XM_017015260.1 | 2 | 215 | XP_016870749.1 | |
| XM_017015261.1 | 3 | 437 | XP_016870750.1 | |
| XM_017015262.1 | 3 | 926 | XP_016870751.1 | |
| XM_017015263.1 | 3 | 341 | XP_016870752.1 | |
| XM_017015264.1 | XP_016870753.1 | |||
| XM_017015265.1 | 1 | 625 | XP_016870754.1 | |
| XM_017015266.1 | 2 | 256 | XP_016870755.1 | |
| XM_017015267.1 | XP_016870756.1 | |||
| XM_017015268.1 | 2 | 239 | XP_016870757.1 | |
| AK094505.1 | 2 | 656 | ||
| AK291178.1 | ||||
| AK297995.1 | 3 | 316 | ||
| AK314427.1 | ||||
| AU123740.1 | ||||
| BC033685.1 | AAH33685.1 | |||
| BI755163.1 | ||||
| DA381449.1 | ||||
| DC413592.1 | 3 | 352 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6724 | Chr.9:128823206 - 128858415 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map