Assay Details
Target Gene Details
Entrez Gene ID: | 5047 |
Gene Name: | progestagen associated endometrial protein |
Gene Aliases: |
GD, GdA, GdF, GdS, PAEG, PEP, PP14 |
Location: |
Chr.9:135561624-135566776 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAEP | NM_001018048.1 | NP_001018058.1 | ||
| NM_001018049.2 | NP_001018059.1 | |||
| NM_002571.3 | NP_002562.2 | |||
| XM_011518745.2 | XP_011517047.1 | |||
| XM_011518746.2 | XP_011517048.1 | |||
| XM_011518747.2 | XP_011517049.1 | |||
| XM_011518748.1 | XP_011517050.1 | |||
| XM_011518749.2 | XP_011517051.1 | |||
| XM_011518751.1 | XP_011517053.1 | |||
| XM_011518752.2 | XP_011517054.1 | |||
| XM_017014782.1 | XP_016870271.1 | |||
| XM_017014783.1 | XP_016870272.1 | |||
| AK304657.1 | ||||
| AK309965.1 | 3 | 516 | ||
| AK309983.1 | ||||
| AK311007.1 | ||||
| AK311730.1 | ||||
| AK311731.1 | 2 | 442 | ||
| AK311813.1 | ||||
| BC069451.1 | AAH69451.1 | |||
| BC069562.1 | AAH69562.1 | |||
| BC112304.1 | AAI12305.1 | |||
| BC113728.1 | ||||
| DC421713.1 | ||||
| DQ884407.1 | ||||
| J04129.1 | AAA60147.1 | |||
| M61886.1 | AAA35802.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv615842 | Chr.9:135113632 - 135571320 on Build GRCh38 | Gain |
|
| nsv615882 | Chr.9:135491367 - 135577889 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map