Assay Details
Target Gene Details
Entrez Gene ID: | 100885775 |
Gene Name: | BRAF-activated non-protein coding RNA |
Gene Aliases: |
LINC00586 |
Location: |
Chr.9:69296681-69307056 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BANCR | NR_047671.2 | |||
| AA442062.1 | ||||
| AA992895.1 | ||||
| AI394221.1 | ||||
| AI472919.1 | ||||
| N98863.1 | ||||
| W58600.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv614548 | Chr.9:68856996 - 69320661 on Build GRCh38 | Gain |
|
| dgv7665n100 | Chr.9:68783797 - 69356369 on Build GRCh38 | Gain |
|
| nsv831610 | Chr.9:69232025 - 69401919 on Build GRCh38 | Gain |
|

Genomic Map