Assay Details
Target Gene Details
Entrez Gene ID: | 27290 |
Gene Name: | serine peptidase inhibitor, Kazal type 4 |
Gene Aliases: |
HEL136, PEC-60, PEC60 |
Location: |
Chr.9:33240198-33248567 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPINK4 | NM_014471.1 | NP_055286.1 | ||
| AF048700.1 | AAC05124.1 | |||
| BC110068.1 | AAI10069.1 | |||
| EU794680.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1023246 | Chr.9:33181636 - 33261063 on Build GRCh38 | Gain |
|
| nsv519369 | Chr.9:33134619 - 33283163 on Build GRCh38 | Gain |
|
| dgv7516n100 | Chr.9:33121909 - 33278846 on Build GRCh38 | Gain |
|
| nsv614146 | Chr.9:33186158 - 33398608 on Build GRCh38 | Gain |
|
| esv2762813 | Chr.9:33139254 - 33261075 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map