Assay Details
Target Gene Details
Entrez Gene ID: | 8727 |
Gene Name: | catenin alpha like 1 |
Gene Aliases: |
ACRP, CLLP, alpha-CATU |
Location: |
Chr.9:108942569-109013594 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTNNAL1 | NM_001286974.1 | NP_001273903.1 | ||
| NM_003798.3 | NP_003789.1 | |||
| XM_005252291.3 | XP_005252348.1 | |||
| XM_017015250.1 | XP_016870739.1 | |||
| XM_017015251.1 | XP_016870740.1 | |||
| AB451283.1 | ||||
| AB451415.1 | ||||
| AF006070.1 | AAC26011.1 | |||
| AF030233.1 | AAC83460.1 | |||
| AF080071.1 | AAC69576.1 | |||
| AF135021.1 | AAF07819.1 | |||
| AK022539.1 | ||||
| AK022834.1 | ||||
| AK022926.1 | ||||
| AK123916.1 | 2 | 1867 | ||
| AK223230.1 | BAD96950.1 | |||
| AK314677.1 | ||||
| AL050016.1 | CAB43238.1 | |||
| BC117208.1 | ||||
| BC117234.1 | ||||
| BU677451.1 | ||||
| U97067.1 | AAC27693.1 |
Target Gene Details
Entrez Gene ID: | 54942 |
Gene Name: | family with sequence similarity 206 member A |
Gene Aliases: |
C9orf6, CG-8, Simiate |
Location: |
Chr.9:108934393-108950745 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM206A | XM_011518816.2 | XP_011517118.1 |
More Information
Set Membership: |
|

Genomic Map