Assay Details
Target Gene Details
Entrez Gene ID: | 54542 |
Gene Name: | ring finger and CCCH-type domains 2 |
Gene Aliases: |
MNAB, RNF164 |
Location: |
Chr.9:122849453-122905283 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RC3H2 | NM_001100588.1 | NP_001094058.1 | ||
| AF255304.1 | AAG00433.1 | |||
| AK027042.1 | BAB15634.1 | |||
| AL833177.1 | CAI46182.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv615270 | Chr.9:122823630 - 122859529 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map