Assay Details
Target Gene Details
Entrez Gene ID: | 100133077 |
Gene Name: | uncharacterized LOC100133077 |
Gene Aliases: |
- |
Location: |
Chr.9:137867925-137892570 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100133077 | NR_121583.1 | |||
| AK128414.1 | BAC87428.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv825200 | Chr.9:137867846 - 137873175 on Build GRCh38 | Gain |
|
| nsv509338 | Chr.9:137839021 - 137984377 on Build GRCh38 | Insertion |
|

Genomic Map