Assay Details
Target Gene Details
Entrez Gene ID: | 138199 |
Gene Name: | carnosine N-methyltransferase 1 |
Gene Aliases: |
C9orf41, UPF0586 |
Location: |
Chr.9:74980794-75028456 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CARNMT1 | NM_001320497.1 | NP_001307426.1 | ||
| NM_152420.2 | NP_689633.1 | |||
| NR_135282.1 | ||||
| XM_005251701.3 | XP_005251758.2 | |||
| XM_017014269.1 | XP_016869758.1 | |||
| XM_017014270.1 | XP_016869759.1 | |||
| XM_017014271.1 | XP_016869760.1 | |||
| AK098661.1 | BAC05369.1 | |||
| AK299691.1 | ||||
| BC034033.1 | AAH34033.1 | |||
| BI560441.1 | ||||
| BX538061.1 | CAD97992.1 | |||
| BX647386.1 |
More Information
Set Membership: |
|

Genomic Map