Assay Details
Target Gene Details
Entrez Gene ID: | 1759 |
Gene Name: | dynamin 1 |
Gene Aliases: |
DNM, EIEE31 |
Location: |
Chr.9:128203355-128255248 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
Target Gene Details
Entrez Gene ID: | 406978 |
Gene Name: | microRNA 199b |
Gene Aliases: |
MIRN199B, mir-199b |
Location: |
Chr.9:128244721-128244830 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR199B | NR_029619.1 | |||
| AJ550412.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951772 | Chr.9:128228322 - 128257921 on Build GRCh38 | Deletion |
|
| nsv831728 | Chr.9:128237994 - 128416514 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map