Assay Details
Target Gene Details
Entrez Gene ID: | 101928550 |
Gene Name: | SMC2 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.9:104080024-104092474 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMC2-AS1 | NR_121580.1 | 3 | 1643 | |
| AK125887.1 | 3 | 1644 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516897 | Chr.9:104051184 - 104088570 on Build GRCh38 | Loss |
|
| dgv7695n100 | Chr.9:104042528 - 104090136 on Build GRCh38 | Loss |
|
| nsv615049 | Chr.9:103527145 - 104134528 on Build GRCh38 | Gain |
|
| nsv831676 | Chr.9:103901194 - 104112176 on Build GRCh38 | Gain |
|
| esv2761294 | Chr.9:104048899 - 104090136 on Build GRCh38 | Loss |
|
| nsv1045887 | Chr.9:104059467 - 104090136 on Build GRCh38 | Loss |
|
| esv3621328 | Chr.9:104050448 - 104090010 on Build GRCh38 | Loss |
|
| nsv615064 | Chr.9:103811066 - 104367820 on Build GRCh38 | Gain |
|

Genomic Map