Assay Details
Target Gene Details
Entrez Gene ID: | 23732 |
Gene Name: | ferric chelate reductase 1 like |
Gene Aliases: |
C9orf4, CG-6, CG6, EIEE37 |
Location: |
Chr.9:109136188-109167291 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FRRS1L | NM_014334.2 | NP_055149.2 | ||
| XM_011518453.2 | XP_011516755.1 | |||
| XM_011518454.2 | XP_011516756.1 | |||
| AF155065.1 | AAF28159.1 | |||
| AI089389.1 |
More Information
Set Membership: |
|

Genomic Map