Assay Details
Target Gene Details
Entrez Gene ID: | 944 |
Gene Name: | tumor necrosis factor superfamily member 8 |
Gene Aliases: |
CD153, CD30L, CD30LG, TNLG3A |
Location: |
Chr.9:114893343-114930595 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNFSF8 | NM_001244.3 | 4 | 2970 | NP_001235.1 |
| NM_001252290.1 | NP_001239219.1 | |||
| BX108999.1 | ||||
| CD690231.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831695 | Chr.9:114805346 - 114963967 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map