Assay Details
Target Gene Details
Entrez Gene ID: | 101929428 |
Gene Name: | PTPRD antisense RNA 2 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.9:10613206-10620420 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTPRD-AS2 | NR_110696.1 | 3 | 2579 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761493 | Chr.9:10617850 - 10627848 on Build GRCh38 | Loss |
|
| nsv831508 | Chr.9:10584843 - 10769475 on Build GRCh38 | Gain |
|
| nsv1032936 | Chr.9:10541177 - 10633543 on Build GRCh38 | Gain |
|
| nsv831507 | Chr.9:10439463 - 10676779 on Build GRCh38 | Loss |
|
| esv3891564 | Chr.9:9553037 - 11021845 on Build GRCh38 | Gain |
|

Genomic Map