Assay Details
Target Gene Details
Entrez Gene ID: | 23245 |
Gene Name: | astrotactin 2 |
Gene Aliases: |
bA67K19.1 |
Location: |
Chr.9:116425225-117415039 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASTN2 | NM_001184734.1 | NP_001171663.1 | ||
| NM_014010.4 | NP_054729.3 | |||
| NM_198186.3 | NP_937829.3 | |||
| NM_198187.3 | NP_937830.3 | |||
| NM_198188.2 | NP_937831.1 | |||
| AB014534.1 | ||||
| AF116574.1 | AAF14357.1 | |||
| AK024064.1 | ||||
| AK127350.1 | ||||
| AK315025.1 | ||||
| BC018759.2 | AAH18759.2 | |||
| BC029272.1 | AAH29272.1 | |||
| BC093835.1 | ||||
| BC101667.1 | ||||
| BC143289.1 | ||||
| BC143292.1 | ||||
| BC143293.1 | ||||
| BC146756.1 | ||||
| BM552790.1 | ||||
| BX649176.1 |
Target Gene Details
Entrez Gene ID: | 100128505 |
Gene Name: | ASTN2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.9:116504283-116562293 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASTN2-AS1 | NR_033973.1 | |||
| AK057447.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831701 | Chr.9:116485758 - 116642973 on Build GRCh38 | Gain |
|
| nsv615254 | Chr.9:116500349 - 116577184 on Build GRCh38 | Loss |
|
| esv3891719 | Chr.9:116525276 - 116552571 on Build GRCh38 | Loss |
|
| esv2761554 | Chr.9:116541782 - 116595630 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map