Assay Details
Target Gene Details
Entrez Gene ID: | 51148 |
Gene Name: | cerebral endothelial cell adhesion molecule |
Gene Aliases: |
CEECAM1, GLT25D3 |
Location: |
Chr.9:128405993-128437351 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CERCAM | NM_001286760.1 | 1 | 41 | NP_001273689.1 |
| XM_005252035.1 | 2 | 539 | XP_005252092.1 | |
| XM_011518762.1 | 2 | 539 | XP_011517064.1 | |
| XM_017014794.1 | 4 | 438 | XP_016870283.1 | |
| AB040935.1 | 2 | 348 | BAA96026.1 | |
| AK314294.1 | 1 | 87 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951774 | Chr.9:128401522 - 128420221 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map