Assay Details
Target Gene Details
Entrez Gene ID: | 144608 |
Gene Name: | chromosome 12 open reading frame 60 |
Gene Aliases: |
- |
Location: |
Chr.12:14567991-14823858 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C12orf60 | XM_017018871.1 | XP_016874360.1 | ||
| XM_017018872.1 | XP_016874361.1 |
Target Gene Details
Entrez Gene ID: | 105369669 |
Gene Name: | uncharacterized LOC105369669 |
Gene Aliases: |
- |
Location: |
Chr.12:14746666-14770662 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105369669 | XM_011520896.2 | XP_011519198.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs74068118] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map