Assay Details
Target Gene Details
Entrez Gene ID: | 3164 |
Gene Name: | nuclear receptor subfamily 4 group A member 1 |
Gene Aliases: |
GFRP1, HMR, N10, NAK-1, NGFIB, NP10, NUR77, TR3 |
Location: |
Chr.12:52022832-52059507 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NR4A1 | NM_001202233.1 | NP_001189162.1 | ||
| NM_001202234.1 | NP_001189163.1 | |||
| XM_005268822.3 | XP_005268879.1 | |||
| XM_006719364.3 | XP_006719427.1 | |||
| XM_017019248.1 | 1 | 2309 | XP_016874737.1 | |
| XM_017019249.1 | 1 | 2309 | XP_016874738.1 | |
| AK131566.1 | BAD18699.1 | |||
| AK297526.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2629n54 | Chr.12:52009326 - 52049869 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map