Assay Details
Target Gene Details
Entrez Gene ID: | 160622 |
Gene Name: | general receptor for phosphoinositides 1 associated scaffold protein |
Gene Aliases: |
TAMALIN |
Location: |
Chr.12:52006945-52015889 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRASP | NM_181711.3 | NP_859062.1 | ||
| XM_005268691.3 | 1 | 448 | XP_005268748.1 | |
| BC053666.1 | AAH53666.1 | |||
| HY052304.1 |
More Information
Set Membership: |
|

Genomic Map