Assay Details
Target Gene Details
Entrez Gene ID: | 29951 |
Gene Name: | PDZ domain containing ring finger 4 |
Gene Aliases: |
LNX4, SAMCAP3L |
Location: |
Chr.12:41188448-41574590 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDZRN4 | NM_001164595.1 | NP_001158067.1 | ||
| AK294535.1 | ||||
| AW297403.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1048258 | Chr.12:39970608 - 41209299 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map