Assay Details
Target Gene Details
Entrez Gene ID: | 51290 |
Gene Name: | ERGIC and golgi 2 |
Gene Aliases: |
CDA14, Erv41, PTX1, cd002 |
Location: |
Chr.12:29336544-29381234 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERGIC2 | AK097112.1 | 1 | 1810 | |
| AL834128.2 | 14 | 4719 | CAD89902.1 |

Genomic Map