Assay Details
Target Gene Details
Entrez Gene ID: | 8739 |
Gene Name: | harakiri, BCL2 interacting protein |
Gene Aliases: |
DP5, HARAKIRI |
Location: |
Chr.12:116856144-116881427 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HRK | NM_003806.2 | NP_003797.1 | ||
| NR_073189.1 | ||||
| XM_017020146.1 | XP_016875635.1 | |||
| BE674890.1 | ||||
| D83699.1 | ||||
| U76376.1 | AAC34931.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv898 | Chr.12:116849419 - 116884372 on Build GRCh38 | Insertion |
|
| nsv832524 | Chr.12:116876305 - 117058771 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map