Assay Details
Target Gene Details
Entrez Gene ID: | 64581 |
Gene Name: | C-type lectin domain family 7 member A |
Gene Aliases: |
BGR, CANDF4, CD369, CLECSF12, DECTIN1, SCARE2 |
Location: |
Chr.12:10116777-10130269 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
More Information
Set Membership: |
|

Genomic Map