Assay Details
Target Gene Details
Entrez Gene ID: | 92293 |
Gene Name: | transmembrane protein 132C |
Gene Aliases: |
PPP1R152 |
Location: |
Chr.12:128267170-128707915 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM132C | NM_001136103.2 | NP_001129575.2 | ||
| XM_011538998.2 | XP_011537300.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1573n100 | Chr.12:126502891 - 128427644 on Build GRCh38 | Gain |
|
| nsv1160070 | Chr.12:128223477 - 128299998 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78942625] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map