Assay Details
Target Gene Details
Entrez Gene ID: | 253827 |
Gene Name: | methionine sulfoxide reductase B3 |
Gene Aliases: |
DFNB74 |
Location: |
Chr.12:65278643-65466907 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MSRB3 | NM_001031679.2 | NP_001026849.1 | ||
| NM_001193460.1 | NP_001180389.1 | |||
| NM_001193461.1 | NP_001180390.1 | |||
| NM_198080.3 | NP_932346.1 | |||
| AK293084.1 | ||||
| AK299065.1 | ||||
| AK316365.1 | ||||
| AL833622.1 | ||||
| AY358229.1 | AAQ88596.1 | |||
| BC040053.1 | AAH40053.1 | |||
| BX500552.1 | ||||
| BX648776.1 | CAI46018.1 | |||
| DC357690.1 |
More Information
Set Membership: |
|

Genomic Map