Assay Details
Target Gene Details
Entrez Gene ID: | 246329 |
Gene Name: | SH3 and cysteine rich domain 3 |
Gene Aliases: |
NAM |
Location: |
Chr.12:57243453-57251193 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STAC3 | NM_001286256.1 | NP_001273185.1 | ||
| NM_001286257.1 | NP_001273186.1 | |||
| NM_145064.2 | NP_659501.1 | |||
| NR_104422.1 | ||||
| XM_011538126.2 | XP_011536428.1 | |||
| AK057013.1 | BAB71343.1 | |||
| AK300688.1 | ||||
| AK311513.1 | ||||
| BC008069.2 | AAH08069.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map