Assay Details
Target Gene Details
Entrez Gene ID: | 8738 |
Gene Name: | CASP2 and RIPK1 domain containing adaptor with death domain |
Gene Aliases: |
MRT34, RAIDD |
Location: |
Chr.12:93677342-93894840 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRADD | NM_001320099.1 | NP_001307028.1 | ||
| NM_001320100.1 | NP_001307029.1 | |||
| NM_001320101.1 | NP_001307030.1 | |||
| NM_003805.4 | NP_003796.1 | |||
| NR_135147.1 | ||||
| XM_017020142.1 | XP_016875631.1 | |||
| XM_017020143.1 | XP_016875632.1 | |||
| XM_017020144.1 | XP_016875633.1 | |||
| AB451280.1 | ||||
| AB451412.1 | ||||
| AI417529.1 | ||||
| AI791547.1 | ||||
| AK294986.1 | ||||
| AK296225.1 | ||||
| AK314861.1 | ||||
| BC017042.1 | AAH17042.1 | |||
| BC037905.1 | AAH37905.1 | |||
| BT009837.1 | AAP88839.1 | |||
| CR407643.1 | CAG28571.1 | |||
| HY113325.1 | ||||
| U79115.1 | AAB42217.1 | |||
| U84388.1 | AAC50952.1 |
Target Gene Details
Entrez Gene ID: | 101928731 |
Gene Name: | uncharacterized LOC101928731 |
Gene Aliases: |
- |
Location: |
Chr.12:93707791-93737823 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928731 | NR_110092.1 | |||
| NR_110093.1 | ||||
| AK130911.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv826 | Chr.12:93729104 - 93774535 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map