Assay Details
Target Gene Details
Entrez Gene ID: | 904 |
Gene Name: | cyclin T1 |
Gene Aliases: |
CCNT, CYCT1, HIVE1 |
Location: |
Chr.12:48688458-48716998 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCNT1 | NM_001240.3 | NP_001231.2 | ||
| NM_001277842.1 | NP_001264771.1 | |||
| XM_017020197.1 | XP_016875686.1 | |||
| AF045161.1 | AAC39638.1 | |||
| AF048730.1 | AAC39664.1 | |||
| AK290990.1 | ||||
| EF688064.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs75063057] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map