Assay Details
Target Gene Details
Entrez Gene ID: | 1368 |
Gene Name: | carboxypeptidase M |
Gene Aliases: |
- |
Location: |
Chr.12:68842200-68963240 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPM |
Target Gene Details
Entrez Gene ID: | 4193 |
Gene Name: | MDM2 proto-oncogene |
Gene Aliases: |
ACTFS, HDMX, hdm2 |
Location: |
Chr.12:68808149-68845544 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MDM2 | NM_001145337.2 | 11 | 5936 | NP_001138809.1 |
| NM_001145339.2 | 9 | 6162 | NP_001138811.1 | |
| NM_001145340.2 | 5 | 5489 | NP_001138812.1 | |
| NM_001278462.1 | 6 | 5567 | NP_001265391.1 | |
| NM_002392.5 | 11 | 6327 | NP_002383.2 | |
| XM_005268872.4 | 11 | 6255 | XP_005268929.1 | |
| XM_006719399.3 | 9 | 5910 | XP_006719462.1 | |
| XM_006719400.3 | 10 | 6216 | XP_006719463.1 |
More Information
Set Membership: |
|

Genomic Map