Assay Details
Target Gene Details
Entrez Gene ID: | 55332 |
Gene Name: | DNA damage regulated autophagy modulator 1 |
Gene Aliases: |
DRAM |
Location: |
Chr.12:101877327-101923623 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DRAM1 | NM_018370.2 | NP_060840.2 | ||
| XM_005269004.2 | XP_005269061.1 | |||
| XM_005269005.2 | XP_005269062.1 | |||
| XM_017019578.1 | XP_016875067.1 | |||
| AK094923.1 | ||||
| AK297785.1 | ||||
| BC018435.1 | AAH18435.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1053046 | Chr.12:101860641 - 102015846 on Build GRCh38 | Gain |
|
| nsv560030 | Chr.12:101805189 - 101901440 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78400917] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map